accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CNHONHNHOSOOOOCH3(Z)

CAS RN 14936-97-1

IUPAC Name: sodium 3-{2-[(1Z)-3-[(2,4-dimethylphenyl)carbamoyl]-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-4-hydroxybenzene-1-sulfonate
Synonyms: sodium 3-{2-[(1Z)-3-[(2,4-dimethylphenyl)carbamoyl]-2-oxonaphthalen-1-ylidene]hydrazin-1-yl}-4-hydroxybenzenesulfonate
Molecular Weight (g/mol): 513.5
Molecular Formula: C25H20N3NaO6S
InChi Key: LPXSXFXFNSMAOI-MCOPDZHSSA-M
SMILES: [Na+].CC1=CC=C(NC(=O)C2=CC3=CC=CC=C3\C(=N\NC3=CC(=CC=C3O)S([O-])(=O)=O)C2=O)C(C)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results