accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNFFF

CAS RN 1494-26-4

IUPAC Name: 4-methoxy-N-[3-(trifluoromethyl)phenyl]aniline
Synonyms: 4-methoxy-N-[3-(trifluoromethyl)phenyl]aniline
Molecular Weight (g/mol): 267.25
Molecular Formula: C14H12F3NO
InChi Key: VODIERGLAGAWQU-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC2=CC=CC(=C2)C(F)(F)F)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results