accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHF

CAS RN 149437-76-3

IUPAC Name: 5-(4-fluorophenyl)-5-oxopentanoic acid
Synonyms: 5-(4-fluorophenyl)-5-oxopentanoic acid
Molecular Weight (g/mol): 210.2
Molecular Formula: C11H11FO3
InChi Key: ZBQROUOOMAMCQW-UHFFFAOYSA-N
SMILES: OC(=O)CCCC(=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results