CAS RN 149488-93-7
IUPAC Name:
2-(2-chloro-6-fluorophenyl)ethan-1-aminium
Synonyms:
2-(2-chloro-6-fluorophenyl)ethanaminium
Molecular Weight (g/mol):
174.62
Molecular Formula:
C8H10ClFN
InChi Key:
NNEMMAAHBKHXLP-UHFFFAOYSA-O
SMILES:
[NH3+]CCC1=C(F)C=CC=C1Cl
1
–
1
of
1
results
1
Promotions Available
2-Chloro-6-fluorophenethylamine, 97%, Thermo Scientific™
2-Chloro-6-fluorophenethylamine, 97%, Thermo Scientific™
CAS: 149488-93-7 Molecular Formula: C8H9ClFN Molecular Weight (g/mol): 173.62 MDL Number: MFCD00052682 InChI Key: NNEMMAAHBKHXLP-UHFFFAOYSA-N PubChem CID: 2734094 IUPAC Name: 2-(2-chloro-6-fluorophenyl)ethanamine SMILES: C1=CC(=C(C(=C1)Cl)CCN)F