accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOCH3

CAS RN 149777-00-4

IUPAC Name: 4,4-dimethyl oxane-4,4-dicarboxylate
Synonyms: 4,4-dimethyl oxane-4,4-dicarboxylate
Molecular Weight (g/mol): 202.21
Molecular Formula: C9H14O5
InChi Key: OTIZXJJKKRZQJR-UHFFFAOYSA-N
SMILES: COC(=O)C1(CCOCC1)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results