accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3COCH3(E/Z)

CAS RN 149777-83-3

IUPAC Name: 2-[2-(4-methoxyphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[2-(4-methoxyphenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 260.14
Molecular Formula: C15H21BO3
InChi Key: VXSHTICWQKLRMP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=CB2OC(C)(C)C(C)(C)O2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results