accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 149777-84-4

IUPAC Name: 4,4,5,5-tetramethyl-2-[2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[2-(4-methylphenyl)ethenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 244.14
Molecular Formula: C15H21BO2
InChi Key: HHBWKASJNTZJLB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=CB2OC(C)(C)C(C)(C)O2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results