accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14979-11-4

IUPAC Name: (3Z,6E)-3,6-bis[2-(4-methyl-2-sulfophenyl)hydrazin-1-ylidene]-4,5-dioxo-3,4,5,6-tetrahydronaphthalene-2,7-disulfonic acid
Synonyms: (3Z,6E)-3,6-bis[2-(4-methyl-2-sulfophenyl)hydrazin-1-ylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
Molecular Weight (g/mol): 716.68
Molecular Formula: C24H20N4O14S4
InChi Key: CAWGMVYQLWPWHZ-KKTFQPMKSA-N
SMILES: CC1=CC=C(N\N=C2\C(=O)C3=C(C=C2S(O)(=O)=O)C=C(\C(=N\NC2=CC=C(C)C=C2S(O)(=O)=O)C3=O)S(O)(=O)=O)C(=C1)S(O)(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results