accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHCH3

CAS RN 14979-39-6

IUPAC Name: 4-methylheptan-3-ol
Synonyms: 4-methylheptan-3-ol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: BKQICAFAUMRYLZ-UHFFFAOYNA-N
SMILES: CCCC(C)C(O)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)