accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 149806-06-4

IUPAC Name: 6-bromopyridine-3-carbaldehyde
Synonyms: 6-bromopyridine-3-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: PVUKGNBRJFTFNJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=O)C=N1
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)