accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 14984-21-5

IUPAC Name: bis(4-phenoxyphenyl)methanone
Synonyms: bis(4-phenoxyphenyl)methanone
Molecular Weight (g/mol): 366.42
Molecular Formula: C25H18O3
InChi Key: BSILAEQTGTZMIW-UHFFFAOYSA-N
SMILES: O=C(C1=CC=C(OC2=CC=CC=C2)C=C1)C1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results