accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOOFSOOFK

CAS RN 14984-76-0

IUPAC Name: potassium bis(fluorosulfonyl)azanide
Synonyms: potassium bis(fluorosulfonyl)azanide
Molecular Weight (g/mol): 219.22
Molecular Formula: F2KNO4S2
InChi Key: MHEBVKPOSBNNAC-UHFFFAOYSA-N
SMILES: [K+].FS(=O)(=O)[N-]S(F)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results