accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14987-04-3

IUPAC Name: dimagnesium(2+) spiro[3.3]trisiloxane-2,2,6,6-tetrakis(olate) hydrate
Synonyms: magnesium trisilicate
Molecular Weight (g/mol): 278.87
Molecular Formula: H2Mg2O9Si3
InChi Key: DGVMNQYBHPSIJS-UHFFFAOYSA-N
SMILES: O.[Mg++].[Mg++].[O-][Si]1([O-])O[Si]2(O1)O[Si]([O-])([O-])O2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results