accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1499-53-2

IUPAC Name: ethyl 2-(phenylformamido)acetate
Synonyms: ethyl 2-(phenylformamido)acetate
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: PTXRQIPIELXJFH-UHFFFAOYSA-N
SMILES: CCOC(=O)CNC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results