accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 149923-10-4

IUPAC Name: (4-chlorophenyl)methanide; chlorozincylium
Synonyms: (4-chlorophenyl)methanide; chlorozincylium
Molecular Weight (g/mol): 226.41
Molecular Formula: C7H6Cl2Zn
InChi Key: GIUOJMHOGPURPS-UHFFFAOYSA-M
SMILES: Cl[Zn+].[CH2-]C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)