accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14996-60-2

IUPAC Name: trichloroosmium hydrate
Synonyms: trichloroosmium hydrate
Molecular Weight (g/mol): 314.6
Molecular Formula: Cl3H2OOs
InChi Key: KHHOLOMEVBPRQY-UHFFFAOYSA-K
SMILES: O.Cl[Os](Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results