CAS RN 150-38-9
CAS RN 150-38-9
IUPAC Name:
trisodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxymethyl)amino)acetate trihydrate
Synonyms:
trisodium edta
Molecular Weight (g/mol):
412.23
Molecular Formula:
C10H19N2Na3O11
InChi Key:
FXNQQEVEDZAAJM-UHFFFAOYSA-K
SMILES:
O.O.O.[Na+].[Na+].[Na+].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ EDTA, 0.5 M, pH 8.0, Molecular Biology Grade, DEPC-Treated
Suitable for use in molecular biology applications