accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150-83-4

IUPAC Name: sodium 3-hydroxybutanoate
Synonyms: sodium 3-hydroxybutyrate
Molecular Weight (g/mol): 126.09
Molecular Formula: C4H7NaO3
InChi Key: NBPUSGBJDWCHKC-UHFFFAOYNA-M
SMILES: [Na+].CC(O)CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)