accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150009-58-8

IUPAC Name: 4-(benzyloxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
Synonyms: 4-(benzyloxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxobutanoic acid
Molecular Weight (g/mol): 445.47
Molecular Formula: C26H23NO6
InChi Key: OQGAELAJEGGNKG-UHFFFAOYNA-N
SMILES: OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)