accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150010-20-1

IUPAC Name: 2-bromo-5-methylpyrimidine
Synonyms: 2-bromo-5-methylpyrimidine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: KYCGEJNZMHUBMX-UHFFFAOYSA-N
SMILES: CC1=CN=C(Br)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results