accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15009-91-3

IUPAC Name: 2-nitropyridine
Synonyms: 2-nitropyridine
Molecular Weight (g/mol): 124.1
Molecular Formula: C5H4N2O2
InChi Key: HLTDBMHJSBSAOM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results