CAS RN 15015-57-3
IUPAC Name:
4-[(4-hydroxyphenyl)disulfanyl]phenol
Synonyms:
4-[(4-hydroxyphenyl)disulfanyl]phenol
Molecular Weight (g/mol):
250.33
Molecular Formula:
C12H10O2S2
InChi Key:
XGKGITBBMXTKTE-UHFFFAOYSA-N
SMILES:
OC1=CC=C(SSC2=CC=C(O)C=C2)C=C1
1
–
1
of
1
results
1
Bis(4-hydroxyphenyl) Disulfide 98.0+%, TCI America™
CAS: 15015-57-3 Molecular Formula: C12H10O2S2 Molecular Weight (g/mol): 250.33 MDL Number: MFCD00020164 InChI Key: XGKGITBBMXTKTE-UHFFFAOYSA-N PubChem CID: 280665 IUPAC Name: 4-[(4-hydroxyphenyl)disulfanyl]phenol SMILES: C1=CC(=CC=C1O)SSC2=CC=C(C=C2)O