CAS RN 15016-42-9
IUPAC Name:
(2-ethenylphenyl)boronic acid
Synonyms:
2-ethenylphenylboronic acid
Molecular Weight (g/mol):
147.97
Molecular Formula:
C8H9BO2
InChi Key:
QHFAXRHEKNHTDH-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=CC=C1C=C
1
–
1
of
1
results