CAS RN 150190-03-7
CAS RN 150190-03-7
IUPAC Name:
ethyl 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylate
Synonyms:
ethyl 2-methyl-6-oxo-1H-pyridine-4-carboxylate
Molecular Weight (g/mol):
181.19
Molecular Formula:
C9H11NO3
InChi Key:
WZUAZSMUJJUHMM-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(=O)NC(C)=C1
1
–
1
of
1
results
1
Ethyl 2-hydroxy-6-methylpyridine-4-carboxylate, 97%
CAS: 150190-03-7 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.191 MDL Number: MFCD09953494 InChI Key: WZUAZSMUJJUHMM-UHFFFAOYSA-N Synonym: ethyl 2-hydroxy-6-methylisonicotinate,ethyl 2-hydroxy-6-methylpyridine-4-carboxylate,ethyl 6-methyl-2-oxo-1,2-dihydropyridine-4-carboxylate,4-pyridinecarboxylicacid, 1,2-dihydro-6-methyl-2-oxo-, ethyl ester,acmc-20edky,2-hydroxy-6-methylpyridine-4-carboxylic acid ethyl ester,2-methyl-6-oxo-1h-pyridine-4-carboxylic acid ethyl ester,ethyl 2-methyl-6-oxidanylidene-1h-pyridine-4-carboxylate PubChem CID: 12089326 IUPAC Name: ethyl 2-methyl-6-oxo-1H-pyridine-4-carboxylate SMILES: CCOC(=O)C1=CC(=O)NC(=C1)C