accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2

CAS RN 1502-03-0

IUPAC Name: cyclododecanamine
Synonyms: cyclododecanamine
Molecular Weight (g/mol): 183.34
Molecular Formula: C12H25N
InChi Key: HBGGBCVEFUPUNY-UHFFFAOYSA-N
SMILES: NC1CCCCCCCCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results