CAS RN 1503-49-7
IUPAC Name:
4-benzoylbenzonitrile
Synonyms:
4-benzoylbenzonitrile
Molecular Weight (g/mol):
207.23
Molecular Formula:
C14H9NO
InChi Key:
YSZWJJANSNFQMM-UHFFFAOYSA-N
SMILES:
O=C(C1=CC=CC=C1)C1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
Promotions Available
4-Cyanobenzophenone, 98%
4-Cyanobenzophenone, 98%
CAS: 1503-49-7 Molecular Formula: C14H9NO Molecular Weight (g/mol): 207.232 MDL Number: MFCD00016382 InChI Key: YSZWJJANSNFQMM-UHFFFAOYSA-N PubChem CID: 73921 IUPAC Name: 4-benzoylbenzonitrile SMILES: C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N