accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOH

CAS RN 15031-78-4

IUPAC Name: 2-(pyridin-4-yl)propan-2-ol
Synonyms: 2-(pyridin-4-yl)propan-2-ol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: WEQVCDIJDUVSID-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results