accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150322-43-3

IUPAC Name: 5-[2-cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4H,5H,6H,7H-thieno[3,2-c]pyridin-2-yl acetate
Synonyms: prasugrel efient effient prasita
Molecular Weight (g/mol): 373.44
Molecular Formula: C20H20FNO3S
InChi Key: DTGLZDAWLRGWQN-UHFFFAOYNA-N
SMILES: CC(=O)OC1=CC2=C(CCN(C2)C(C(=O)C2CC2)C2=CC=CC=C2F)S1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)