accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COONCH3NH2

CAS RN 150349-36-3

IUPAC Name: tert-butyl N-(3-aminopropyl)-N-methylcarbamate
Synonyms: tert-butyl N-(3-aminopropyl)-N-methylcarbamate
Molecular Weight (g/mol): 188.27
Molecular Formula: C9H20N2O2
InChi Key: PNQYAMWGTGWJDW-UHFFFAOYSA-N
SMILES: CN(CCCN)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)