CAS RN 1504-41-2
IUPAC Name:
2-(2-aminoacetamido)hexanoic acid
Synonyms:
2-(2-aminoacetamido)hexanoic acid
Molecular Weight (g/mol):
188.23
Molecular Formula:
C8H16N2O3
InChi Key:
XVUIZOUTLADVIH-UHFFFAOYNA-N
SMILES:
CCCCC(NC(=O)CN)C(O)=O
1
–
1
of
1
results
1
Glycyl-DL-norleucine 98.0+%, TCI America™
CAS: 1504-41-2 Molecular Formula: C8H16N2O3 Molecular Weight (g/mol): 188.227 MDL Number: MFCD00038187 InChI Key: XVUIZOUTLADVIH-UHFFFAOYSA-N Synonym: H-Gly-DL-Nle-OH PubChem CID: 273080 IUPAC Name: 2-[(2-aminoacetyl)amino]hexanoic acid SMILES: CCCCC(C(=O)O)NC(=O)CN