accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOOOHOHOHOOHHOOH

CAS RN 150412-80-9

IUPAC Name: 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 448.42
Molecular Formula: C19H28O12
InChi Key: PJFQMORRZLJWQE-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(CO)C(OC3OC(CO)C(O)C(O)C3O)C(O)C2O)C=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)