accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15058-72-7

IUPAC Name: 2,4-dimethylfuran-3-carboxylic acid
Synonyms: 2,4-dimethylfuran-3-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: JFXXYKVSLHRBAA-UHFFFAOYSA-N
SMILES: CC1=C(C(O)=O)C(C)=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results