accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1506-53-2

IUPAC Name: N-dodecylprop-2-enamide
Synonyms: N-dodecylprop-2-enamide
Molecular Weight (g/mol): 239.4
Molecular Formula: C15H29NO
InChi Key: XQPVIMDDIXCFFS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCNC(=O)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results