accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOCl

CAS RN 15068-35-6

IUPAC Name: 2-chloro-3-methylbenzoic acid
Synonyms: 2-chloro-3-methylbenzoic acid
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: LWOKLXMNGXXORN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results