accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150763-46-5

H3CO

CAS RN 150763-46-5

IUPAC Name: 4'-ethyl-[1,1'-bi(cyclohexane)]-4-one
Synonyms: 4'-ethyl-[1,1'-bi(cyclohexane)]-4-one
Molecular Weight (g/mol): 208.35
Molecular Formula: C14H24O
InChi Key: ICCIOWLZPJIEBA-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1CCC(=O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results