CAS RN 1508-65-2
IUPAC Name:
4-(diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride
Synonyms:
oxybutynin hydrochloride
Molecular Weight (g/mol):
393.95
Molecular Formula:
C22H32ClNO3
InChi Key:
SWIJYDAEGSIQPZ-UHFFFAOYNA-N
SMILES:
Cl.CCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)C1=CC=CC=C1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Oxybutynin hydrochloride
CAS: 1508-65-2 Molecular Weight (g/mol): 393.95 InChI Key: SWIJYDAEGSIQPZ-UHFFFAOYNA-N IUPAC Name: 4-(diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride SMILES: Cl.CCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)C1=CC=CC=C1
2
Medisca Oxybutynin Chloride USP
Oxybutynin Chloride USP is a white crystalline powder, which is practically odorless . Alternative name(s): Oxybutynin hydrochloride