accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15082-28-7

IUPAC Name: 2-{[1,1'-biphenyl]-4-yl}-5-(4-tert-butylphenyl)-1,3,4-oxadiazole
Synonyms: butyl pbd
Molecular Weight (g/mol): 354.45
Molecular Formula: C24H22N2O
InChi Key: XZCJVWCMJYNSQO-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=NN=C(O1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)