accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 150828-96-9

CH3H3CCH3OONHHNOOOOH

CAS RN 150828-96-9

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-5-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-5-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 454.52
Molecular Formula: C25H30N2O6
InChi Key: YEBWACZYMHWWEK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)