accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15084-51-2

IUPAC Name: 4-tert-butylbenzene-1-sulfonyl chloride
Synonyms: 4-tert-butylbenzenesulfonyl chloride
Molecular Weight (g/mol): 232.72
Molecular Formula: C10H13ClO2S
InChi Key: YEZADZMMVHWFIY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)