accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONO(S)N

CAS RN 150884-56-3

IUPAC Name: tert-butyl (3S)-3-(4-cyanophenyl)oxaziridine-2-carboxylate
Synonyms: tert-butyl (3S)-3-(4-cyanophenyl)oxaziridine-2-carboxylate
Molecular Weight (g/mol): 246.27
Molecular Formula: C13H14N2O3
InChi Key: ACXPNVRTMHEHMQ-VPHXOMNUSA-N
SMILES: CC(C)(C)OC(=O)N1O[C@H]1C1=CC=C(C=C1)C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results