accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15097-38-8

IUPAC Name: benzyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Synonyms: benzyl 2-(triphenyl-λ⁵-phosphanylidene)acetate
Molecular Weight (g/mol): 410.45
Molecular Formula: C27H23O2P
InChi Key: INKMLGJBBDRIQR-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results