accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 151-56-4

IUPAC Name: aziridine
Synonyms: aziridine rcra waste number P054 pei-14M aziridine, conjugate acid ethylenimine pei-7 conjugate acid aziridine aethylenimin ethylenimine (substance) ccris 296
Molecular Weight (g/mol): 43.07
Molecular Formula: C2H5N
InChi Key: NOWKCMXCCJGMRR-UHFFFAOYSA-N
SMILES: C1CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results