accessibility menu, dialog, popup

UserName

Viewing profile as user:
OClHNOOH

CAS RN 151057-13-5

IUPAC Name: 7-chloro-3-cyclopropanecarbonyl-4-hydroxy-1,2-dihydroquinolin-2-one
Synonyms: 7-chloro-3-cyclopropanecarbonyl-4-hydroxy-1H-quinolin-2-one
Molecular Weight (g/mol): 263.68
Molecular Formula: C13H10ClNO3
InChi Key: MXEFWCFPCLDOOG-UHFFFAOYSA-N
SMILES: OC1=C(C(=O)C2CC2)C(=O)NC2=CC(Cl)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results