accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 151059-43-7

IUPAC Name: 2-phenylethan-1-amine hydroiodide
Synonyms: 2-phenylethylamine hydroiodide
Molecular Weight (g/mol): 249.1
Molecular Formula: C8H12IN
InChi Key: UPHCENSIMPJEIS-UHFFFAOYSA-N
SMILES: I.NCCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)