accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNOH3CNOOOHFH2OH2O

CAS RN 151060-21-8

IUPAC Name: 1-cyclopropyl-6-fluoro-8-methoxy-7-[3-(methylamino)piperidin-1-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic acid dihydrate
Synonyms: balofloxacin dihydrate
Molecular Weight (g/mol): 425.46
Molecular Formula: C20H28FN3O6
InChi Key: WEGNYJXOCPJAPS-UHFFFAOYNA-N
SMILES: O.O.CNC1CCCN(C1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1OC)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results