accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOH

CAS RN 151155-53-2

IUPAC Name: (2,3-dihydro-1-benzofuran-7-yl)methanol
Synonyms: 2,3-dihydro-1-benzofuran-7-ylmethanol
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: WUXXIPOWZJYRNE-UHFFFAOYSA-N
SMILES: OCC1=CC=CC2=C1OCC2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results