accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 151169-67-4

IUPAC Name: (4-chloro-3-nitrophenyl)boronic acid
Synonyms: 4-chloro-3-nitrophenylboronic acid
Molecular Weight (g/mol): 201.37
Molecular Formula: C6H5BClNO4
InChi Key: LQCCRPUZTXKDGB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results