accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNNHNI

CAS RN 151266-23-8

IUPAC Name: 3-iodo-2H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms: 3-iodo-2H-pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol): 261.03
Molecular Formula: C5H4IN5
InChi Key: HQAIUXZORKJOJY-UHFFFAOYSA-N
SMILES: NC1=NC=NC2=NNC(I)=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)