accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNOO

CAS RN 15128-82-2

IUPAC Name: 2-nitropyridin-3-ol
Synonyms: 3-pyridinol, 2-nitro-
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: QBPDSKPWYWIHGA-UHFFFAOYSA-N
SMILES: OC1=CC=CN=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)