accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFFFFFOOH3C

CAS RN 1513-62-8

IUPAC Name: 1,5-dimethyl 2,2,3,3,4,4-hexafluoropentanedioate
Synonyms: 1,5-dimethyl 2,2,3,3,4,4-hexafluoropentanedioate
Molecular Weight (g/mol): 268.11
Molecular Formula: C7H6F6O4
InChi Key: PJZVBCPDYSZAJU-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results